4-Bromo-3,3,4,4-tetrafluorobutyl 4,4,5,5,5-pentafluoropentyl carbonate

Lieferant: Apollo Scientific

PC450392-25G
APOSPC450392-25GEA 1540 EUR
APOSPC450392-25G
4-Bromo-3,3,4,4-tetrafluorobutyl 4,4,5,5,5-pentafluoropentyl carbonate
4-Bromo-3,3,4,4-tetrafluorobutyl 4,4,5,5,5-pentafluoropentyl carbonate
Formel: C₁₀H₁₀BrF₉O₃
Molecular Weight: 429,074 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334464
CAS-Nummer: 1923267-08-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR