4-Bromo-3,3,4,4-tetrafluorobutyl 1H,1H-perfluorohexyl carbonate
Lieferant: Apollo Scientific
PC450393-25G
APOSPC450393-25GEA
1540
EUR
APOSPC450393-25G
4-Bromo-3,3,4,4-tetrafluorobutyl 1H,1H-perfluorohexyl carbonate
4-Bromo-3,3,4,4-tetrafluorobutyl 1H,1H-perfluorohexyl carbonate
Formel:
C₁₁H₆BrF₁₅O₃ Molecular Weight: 551,044 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28334465 CAS-Nummer: 1980035-06-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...