4-Bromo-3,3,4,4-tetrafluorobutyl 1H,1H,2H,2H-perfluorohexyl carbonate

Lieferant: Apollo Scientific

PC450394-25G
APOSPC450394-25GEA 1540 EUR
APOSPC450394-25G
4-Bromo-3,3,4,4-tetrafluorobutyl 1H,1H,2H,2H-perfluorohexyl carbonate
4-Bromo-3,3,4,4-tetrafluorobutyl 1H,1H,2H,2H-perfluorohexyl carbonate
Formel: C₁₁H₈BrF₁₃O₃
Molecular Weight: 515,063 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334466
CAS-Nummer: 1980035-10-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR