4-Bromo-3,3,4,4-tetrafluorobutyl 1H,1H,7H-perfluoroheptyl carbonate

Lieferant: Apollo Scientific

PC450395-25G
APOSPC450395-25GEA 1540 EUR
APOSPC450395-25G
4-Bromo-3,3,4,4-tetrafluorobutyl 1H,1H,7H-perfluoroheptyl carbonate
4-Bromo-3,3,4,4-tetrafluorobutyl 1H,1H,7H-perfluoroheptyl carbonate
Formel: C₁₂H₇BrF₁₆O₃
Molecular Weight: 583,061 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334467
CAS-Nummer: 1980035-16-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR