4-Bromo-3,3,4,4-tetrafluorobutyl 1H,1H,9H-perfluorononyl carbonate
Lieferant: Apollo Scientific
PC450396-25G
APOSPC450396-25GEA
1540
EUR
APOSPC450396-25G
4-Bromo-3,3,4,4-tetrafluorobutyl 1H,1H,9H-perfluorononyl carbonate
4-Bromo-3,3,4,4-tetrafluorobutyl 1H,1H,9H-perfluorononyl carbonate
Formel:
C₁₄H₇BrF₂₀O₃ Molecular Weight: 683,076 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28334468 CAS-Nummer: 1923267-11-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...