Methyl 2,2,3,3,3-pentafluoropropyl carbonate

Lieferant: Apollo Scientific

PC450397-25G
APOSPC450397-25GEA 1540 EUR
APOSPC450397-25G
Methyl 2,2,3,3,3-pentafluoropropyl carbonate
Methyl 2,2,3,3,3-pentafluoropropyl carbonate
Formel: C₅H₅F₅O₃
Molecular Weight: 208.083 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334375
CAS-Nummer: 156783-97-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR