3-Fluoropropyl methyl carbonate

Lieferant: Apollo Scientific

PC450398-25G
APOSPC450398-25GEA 1540 EUR
APOSPC450398-25G
3-Fluoropropyl methyl carbonate
3-Fluoropropyl methyl carbonate
Formel: C₅H₉FO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD28334376
CAS-Nummer: 872353-07-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR