2,2,3,3,4,4,4-Heptafluorobutyl methyl carbonate

Lieferant: Apollo Scientific

PC450399-25G
APOSPC450399-25GEA 1540 EUR
APOSPC450399-25G
2,2,3,3,4,4,4-Heptafluorobutyl methyl carbonate
2,2,3,3,4,4,4-Heptafluorobutyl methyl carbonate
Formel: C₆H₅F₇O₃
Molecular Weight: 258,091 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334377
CAS-Nummer: 277332-95-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR