2,2,3,4,4,4-Hexafluorobutyl methyl carbonate

Lieferant: Apollo Scientific

PC450400-25G
APOSPC450400-25GEA 1540 EUR
APOSPC450400-25G
2,2,3,4,4,4-Hexafluorobutyl methyl carbonate
2,2,3,4,4,4-Hexafluorobutyl methyl carbonate
Formel: C₆H₆F₆O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD28334378
CAS-Nummer: 106538-79-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR