Methyl 4,4,5,5,5-pentafluoropentyl carbonate
Lieferant: Apollo Scientific
PC450401-25G
APOSPC450401-25GEA
1540
EUR
APOSPC450401-25G
Methyl 4,4,5,5,5-pentafluoropentyl carbonate
Methyl 4,4,5,5,5-pentafluoropentyl carbonate
Formel:
C₇H₉F₅O₃ Molecular Weight: 236,137 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28334379 CAS-Nummer: 1980086-58-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...