Methyl 1H,1H-perfluorohexyl carbonate
Lieferant: Apollo Scientific
PC450402-25G
APOSPC450402-25GEA
1540
EUR
APOSPC450402-25G
Methyl 1H,1H-perfluorohexyl carbonate
Methyl 1H,1H-perfluorohexyl carbonate
Formel:
C₈H₅F₁₁O₃ Molecular Weight: 358,106 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28334380 CAS-Nummer: 1980034-02-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...