Methyl 1H,1H,2H,2H-perfluorohexyl carbonate

Lieferant: Apollo Scientific

PC450403-25G
APOSPC450403-25GEA 1540 EUR
APOSPC450403-25G
Methyl 1H,1H,2H,2H-perfluorohexyl carbonate
Methyl 1H,1H,2H,2H-perfluorohexyl carbonate
Formel: C₈H₇F₉O₃
Molecular Weight: 322,125 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334381
CAS-Nummer: 1980076-59-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR