1H,1H,7H-Perfluorohexyl methyl carbonate

Lieferant: Apollo Scientific

PC450404-25G
APOSPC450404-25GEA 1540 EUR
APOSPC450404-25G
1H,1H,7H-Perfluorohexyl methyl carbonate
1H,1H,7H-Perfluorohexyl methyl carbonate
Formel: C₉H₆F₁₂O₃
Molecular Weight: 390,123 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334382
CAS-Nummer: 1980034-31-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR