1H,1H,9H-Perfluorononyl methyl carbonate
Lieferant: Apollo Scientific
PC450405-25G
APOSPC450405-25GEA
1540
EUR
APOSPC450405-25G
1H,1H,9H-Perfluorononyl methyl carbonate
1H,1H,9H-Perfluorononyl methyl carbonate
Formel:
C₁₁H₆F₁₆O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD28334383 CAS-Nummer: 1980085-40-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...