4-Bromo-3,3,4,4-tetrafluorobutyl methyl carbonate

Lieferant: Apollo Scientific

PC450406-25G
APOSPC450406-25GEA 1540 EUR
APOSPC450406-25G
4-Bromo-3,3,4,4-tetrafluorobutyl methyl carbonate
4-Bromo-3,3,4,4-tetrafluorobutyl methyl carbonate
Formel: C₆H₇BrF₄O₃
Molecular Weight: 283,016 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334469
CAS-Nummer: 1923065-47-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR