Ethyl 3-fluoropropyl carbonate

Lieferant: Apollo Scientific

PC450409-25G
APOSPC450409-25GEA 1540 EUR
APOSPC450409-25G
Ethyl 3-fluoropropyl carbonate
Ethyl 3-fluoropropyl carbonate
Formel: C₆H₁₁FO₃
Molecular Weight: 150,148 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334386
CAS-Nummer: 888974-56-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR