Ethyl 2,2,3,3,4,4,4-heptafluorobutyl carbonate

Lieferant: Apollo Scientific

PC450410-25G
APOSPC450410-25GEA 1540 EUR
APOSPC450410-25G
Ethyl 2,2,3,3,4,4,4-heptafluorobutyl carbonate
Ethyl 2,2,3,3,4,4,4-heptafluorobutyl carbonate
Formel: C₇H₇F₇O₃
Molecular Weight: 272,118 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334387
CAS-Nummer: 1980034-03-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR