Ethyl 2,2,3,4,4,4-hexafluorobutyl carbonate

Lieferant: Apollo Scientific

PC450411-25G
APOSPC450411-25GEA 1540 EUR
APOSPC450411-25G
Ethyl 2,2,3,4,4,4-hexafluorobutyl carbonate
Ethyl 2,2,3,4,4,4-hexafluorobutyl carbonate
Formel: C₇H₈F₆O₃
Molecular Weight: 254,127 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334388
CAS-Nummer: 1035454-56-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR