Ethyl 4,4,5,5,5-pentafluoropentyl carbonate

Lieferant: Apollo Scientific

PC450412-25G
APOSPC450412-25GEA 1540 EUR
APOSPC450412-25G
Ethyl 4,4,5,5,5-pentafluoropentyl carbonate
Ethyl 4,4,5,5,5-pentafluoropentyl carbonate
Formel: C₈H₁₁F₅O₃
Molecular Weight: 250,163 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334389
CAS-Nummer: 1980034-40-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR