Ethyl 1H,1H-perfluorohexyl carbonate
Lieferant: Apollo Scientific
PC450413-25G
APOSPC450413-25GEA
1540
EUR
APOSPC450413-25G
Ethyl 1H,1H-perfluorohexyl carbonate
Ethyl 1H,1H-perfluorohexyl carbonate
Formel:
C₉H₇F₁₁O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD28334390 CAS-Nummer: 1980034-41-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...