Ethyl 1H,1H,2H,2H-perfluorohexyl carbonate

Lieferant: Apollo Scientific

PC450414-25G
APOSPC450414-25GEA 1540 EUR
APOSPC450414-25G
Ethyl 1H,1H,2H,2H-perfluorohexyl carbonate
Ethyl 1H,1H,2H,2H-perfluorohexyl carbonate
Formel: C₉H₉F₉O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD28334391
CAS-Nummer: 1980035-62-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR