1H,1H,7H-Perfluorohexyl ethyl carbonate

Lieferant: Apollo Scientific

PC450415-25G
APOSPC450415-25GEA 1540 EUR
APOSPC450415-25G
1H,1H,7H-Perfluorohexyl ethyl carbonate
1H,1H,7H-Perfluorohexyl ethyl carbonate
Formel: C₁₀H₈F₁₂O₃
Molecular Weight: 404,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334392
CAS-Nummer: 1980034-47-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR