Ethyl 1H,1H,9H-perfluorononyl carbonate

Lieferant: Apollo Scientific

PC450416-25G
APOSPC450416-25GEA 1540 EUR
APOSPC450416-25G
Ethyl 1H,1H,9H-perfluorononyl carbonate
Ethyl 1H,1H,9H-perfluorononyl carbonate
Formel: C₁₂H₈F₁₆O₃
Molecular Weight: 504,165 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334393
CAS-Nummer: 1980086-60-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR