2,2,3,3,3-Pentafluoropropyl propyl carbonate

Lieferant: Apollo Scientific

PC450420-25G
APOSPC450420-25GEA 1540 EUR
APOSPC450420-25G
2,2,3,3,3-Pentafluoropropyl propyl carbonate
2,2,3,3,3-Pentafluoropropyl propyl carbonate
Formel: C₇H₉F₅O₃
Molecular Weight: 236,137 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334396
CAS-Nummer: 1980085-61-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR