Propyl 2,2,3,3-tetrafluoropropyl carbonate

Lieferant: Apollo Scientific

PC450421-25G
APOSPC450421-25GEA 1540 EUR
APOSPC450421-25G
Propyl 2,2,3,3-tetrafluoropropyl carbonate
Propyl 2,2,3,3-tetrafluoropropyl carbonate
Formel: C₇H₁₀F₄O₃
Molecular Weight: 218,146 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334397
CAS-Nummer: 879496-52-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR