3-Fluoropropyl propyl carbonate
Lieferant: Apollo Scientific
PC450422-25G
APOSPC450422-25GEA
1540
EUR
APOSPC450422-25G
3-Fluoropropyl propyl carbonate
3-Fluoropropyl propyl carbonate
Formel:
C₇H₁₃FO₃ Molecular Weight: 164,175 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28334398 CAS-Nummer: 1980039-80-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...