2,2,3,3,4,4,4-Heptafluorobutyl propyl carbonate

Lieferant: Apollo Scientific

PC450423-25G
APOSPC450423-25GEA 1540 EUR
APOSPC450423-25G
2,2,3,3,4,4,4-Heptafluorobutyl propyl carbonate
2,2,3,3,4,4,4-Heptafluorobutyl propyl carbonate
Formel: C₈H₉F₇O₃
Molecular Weight: 286,144 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334399
CAS-Nummer: 1980038-87-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR