2,2,3,4,4,4-Hexafluorobutyl propyl carbonate
Lieferant: Apollo Scientific
PC450424-25G
APOSPC450424-25GEA
1540
EUR
APOSPC450424-25G
2,2,3,4,4,4-Hexafluorobutyl propyl carbonate
2,2,3,4,4,4-Hexafluorobutyl propyl carbonate
Formel:
C₈H₁₀F₆O₃ Molecular Weight: 268,154 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28334400 CAS-Nummer: 1980034-53-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...