4,4,5,5,5-Pentafluoropentyl propyl carbonate

Lieferant: Apollo Scientific

PC450425-25G
APOSPC450425-25GEA 1540 EUR
APOSPC450425-25G
4,4,5,5,5-Pentafluoropentyl propyl carbonate
4,4,5,5,5-Pentafluoropentyl propyl carbonate
Formel: C₉H₁₃F₅O₃
Molecular Weight: 264,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334401
CAS-Nummer: 1980086-71-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR