Propyl 1H,1H-perfluorohexyl carbonate

Lieferant: Apollo Scientific

PC450426-25G
APOSPC450426-25GEA 1540 EUR
APOSPC450426-25G
Propyl 1H,1H-perfluorohexyl carbonate
Propyl 1H,1H-perfluorohexyl carbonate
Formel: C₁₀H₉F₁₁O₃
Molecular Weight: 386,159 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334402
CAS-Nummer: 1980085-42-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR