1H,1H,2H,2H-Perfluorohexyl propyl carbonate

Lieferant: Apollo Scientific

PC450427-25G
APOSPC450427-25GEA 1540 EUR
APOSPC450427-25G
1H,1H,2H,2H-Perfluorohexyl propyl carbonate
1H,1H,2H,2H-Perfluorohexyl propyl carbonate
Formel: C₁₀H₁₁F₉O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD28334403
CAS-Nummer: 1980034-59-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR