1H,1H,7H-Perfluorohexyl propyl carbonate

Lieferant: Apollo Scientific

PC450428-25G
APOSPC450428-25GEA 1540 EUR
APOSPC450428-25G
1H,1H,7H-Perfluorohexyl propyl carbonate
1H,1H,7H-Perfluorohexyl propyl carbonate
Formel: C₁₁H₁₀F₁₂O₃
Molecular Weight: 418,176 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334404
CAS-Nummer: 1980086-59-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR