1H,1H,9H-Perfluorononyl propyl carbonate

Lieferant: Apollo Scientific

PC450429-25G
APOSPC450429-25GEA 1540 EUR
APOSPC450429-25G
1H,1H,9H-Perfluorononyl propyl carbonate
1H,1H,9H-Perfluorononyl propyl carbonate
Formel: C₁₃H₁₀F₁₆O₃
Molecular Weight: 518.191 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334405
CAS-Nummer: 1980039-83-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR