4-Bromo-3,3,4,4-tetrafluorobutyl propyl carbonate

Lieferant: Apollo Scientific

PC450430-25G
APOSPC450430-25GEA 1540 EUR
APOSPC450430-25G
4-Bromo-3,3,4,4-tetrafluorobutyl propyl carbonate
4-Bromo-3,3,4,4-tetrafluorobutyl propyl carbonate
Formel: C₈H₁₁BrF₄O₃
Molecular Weight: 311,069 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334471
CAS-Nummer: 1923267-13-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR