Hexafluoroisopropyl propyl carbonate

Lieferant: Apollo Scientific

PC450431-25G
APOSPC450431-25GEA 1540 EUR
APOSPC450431-25G
Hexafluoroisopropyl propyl carbonate
Hexafluoroisopropyl propyl carbonate
Formel: C₇H₈F₆O₃
Molecular Weight: 254,127 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334406
CAS-Nummer: 1980034-68-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR