Isopropyl 2,2,2-trifluoroethyl carbonate

Lieferant: Apollo Scientific

PC450432-25G
APOSPC450432-25GEA 1540 EUR
APOSPC450432-25G
Isopropyl 2,2,2-trifluoroethyl carbonate
Isopropyl 2,2,2-trifluoroethyl carbonate
Formel: C₆H₉F₃O₃
Molecular Weight: 186,129 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334407
CAS-Nummer: 1860179-11-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR