Isopropyl 2,2,3,3-tetrafluoropropyl carbonate

Lieferant: Apollo Scientific

PC450434-25G
APOSPC450434-25GEA 1540 EUR
APOSPC450434-25G
Isopropyl 2,2,3,3-tetrafluoropropyl carbonate
Isopropyl 2,2,3,3-tetrafluoropropyl carbonate
Formel: C₇H₁₀F₄O₃
Molecular Weight: 218,146 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334409
CAS-Nummer: 1980035-67-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR