3-Fluoropropyl isopropyl carbonate

Lieferant: Apollo Scientific

PC450435-25G
APOSPC450435-25GEA 1540 EUR
APOSPC450435-25G
3-Fluoropropyl isopropyl carbonate
3-Fluoropropyl isopropyl carbonate
Formel: C₇H₁₃FO₃
Molecular Weight: 164,175 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334410
CAS-Nummer: 1980085-44-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR