2,2,3,3,4,4,4-Heptafluorobutyl isopropyl carbonate

Lieferant: Apollo Scientific

PC450436-25G
APOSPC450436-25GEA 1540 EUR
APOSPC450436-25G
2,2,3,3,4,4,4-Heptafluorobutyl isopropyl carbonate
2,2,3,3,4,4,4-Heptafluorobutyl isopropyl carbonate
Formel: C₈H₉F₇O₃
Molecular Weight: 286,144 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334411
CAS-Nummer: 1923065-19-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR