2,2,3,4,4,4-Hexafluorobutyl isopropyl carbonate

Lieferant: Apollo Scientific

PC450437-25G
APOSPC450437-25GEA 1540 EUR
APOSPC450437-25G
2,2,3,4,4,4-Hexafluorobutyl isopropyl carbonate
2,2,3,4,4,4-Hexafluorobutyl isopropyl carbonate
Formel: C₈H₁₀F₆O₃
Molecular Weight: 268,154 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334412
CAS-Nummer: 1980086-61-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR