1H,1H,2H,2H-Perfluorohexyl isopropyl carbonate

Lieferant: Apollo Scientific

PC450440-25G
APOSPC450440-25GEA 1540 EUR
APOSPC450440-25G
1H,1H,2H,2H-Perfluorohexyl isopropyl carbonate
1H,1H,2H,2H-Perfluorohexyl isopropyl carbonate
Formel: C₁₀H₁₁F₉O₃
Molecular Weight: 350,178 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334415
CAS-Nummer: 1980086-72-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR