1H,1H,7H-Perfluorohexyl isopropyl carbonate

Lieferant: Apollo Scientific

PC450441-25G
APOSPC450441-25GEA 1540 EUR
APOSPC450441-25G
1H,1H,7H-Perfluorohexyl isopropyl carbonate
1H,1H,7H-Perfluorohexyl isopropyl carbonate
Formel: C₁₁H₁₀F₁₂O₃
Molecular Weight: 418.176 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334416
CAS-Nummer: 1980076-66-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR