1H,1H,9H-Perfluorononyl isopropyl carbonate

Lieferant: Apollo Scientific

PC450442-25G
APOSPC450442-25GEA 1540 EUR
APOSPC450442-25G
1H,1H,9H-Perfluorononyl isopropyl carbonate
1H,1H,9H-Perfluorononyl isopropyl carbonate
Formel: C₁₃H₁₀F₁₆O₃
Molecular Weight: 518,191 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334417
CAS-Nummer: 1980034-98-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR