4-Bromo-3,3,4,4-tetrafluorobutyl isopropyl carbonate

Lieferant: Apollo Scientific

PC450443-25G
APOSPC450443-25GEA 1540 EUR
APOSPC450443-25G
4-Bromo-3,3,4,4-tetrafluorobutyl isopropyl carbonate
4-Bromo-3,3,4,4-tetrafluorobutyl isopropyl carbonate
Formel: C₈H₁₁BrF₄O₃
Molecular Weight: 311,069 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334472
CAS-Nummer: 1980035-20-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR