Hexafluoroisopropyl isopropyl carbonate

Lieferant: Apollo Scientific

PC450444-25G
APOSPC450444-25GEA 1540 EUR
APOSPC450444-25G
Hexafluoroisopropyl isopropyl carbonate
Hexafluoroisopropyl isopropyl carbonate
Formel: C₇H₈F₆O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD28334418
CAS-Nummer: 1980039-90-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR