Butyl 2,2,2-trifluoroethyl carbonate
Lieferant: Apollo Scientific
PC450445-25G
APOSPC450445-25GEA
1540
EUR
APOSPC450445-25G
Butyl 2,2,2-trifluoroethyl carbonate
Butyl 2,2,2-trifluoroethyl carbonate
Formel:
C₇H₁₁F₃O₃ Molecular Weight: 200,156 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28334419 CAS-Nummer: 482576-30-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...