Butyl 2,2,3,3-tetrafluoropropyl carbonate

Lieferant: Apollo Scientific

PC450447-25G
APOSPC450447-25GEA 1540 EUR
APOSPC450447-25G
Butyl 2,2,3,3-tetrafluoropropyl carbonate
Butyl 2,2,3,3-tetrafluoropropyl carbonate
Formel: C₈H₁₂F₄O₃
Molecular Weight: 232,173 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334421
CAS-Nummer: 1980034-56-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR