Butyl 3-fluoropropyl carbonate

Lieferant: Apollo Scientific

PC450448-25G
APOSPC450448-25GEA 1540 EUR
APOSPC450448-25G
Butyl 3-fluoropropyl carbonate
Butyl 3-fluoropropyl carbonate
Formel: C₈H₁₅FO₃
Molecular Weight: 178,201 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334422
CAS-Nummer: 1980085-63-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR