Butyl 2,2,3,3,4,4,4-heptafluorobutyl carbonate
Lieferant: Apollo Scientific
PC450449-25G
APOSPC450449-25GEA
1540
EUR
APOSPC450449-25G
Butyl 2,2,3,3,4,4,4-heptafluorobutyl carbonate
Butyl 2,2,3,3,4,4,4-heptafluorobutyl carbonate
Formel:
C₉H₁₁F₇O₃ Molecular Weight: 300,171 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28334423 CAS-Nummer: 1980038-95-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...