Butyl 4,4,5,5,5-pentafluoropentyl carbonate

Lieferant: Apollo Scientific

PC450451-25G
APOSPC450451-25GEA 1540 EUR
APOSPC450451-25G
Butyl 4,4,5,5,5-pentafluoropentyl carbonate
Butyl 4,4,5,5,5-pentafluoropentyl carbonate
Formel: C₁₀H₁₅F₅O₃
Molecular Weight: 278,216 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334425
CAS-Nummer: 1980085-48-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR